Tisk…

(2-Methyl-5-(trifluoromethyl)phenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

PC302699-1G PC302699-5G PC302699-10G
APOSPC302699-1GEA 5200 CZK
APOSPC302699-1G APOSPC302699-5G APOSPC302699-10G
(2-Methyl-5-(trifluoromethyl)phenyl)acetic acid 98%
(2-Methyl-5-(trifluoromethyl)phenyl)acetic acid
Vzorec: C₁₀H₉F₃O₂
MW: 218,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832286
CAS číslo: 1000517-01-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR