Tisk…

2-Methyl-5-(trifluoromethyl)phenol 98%

Dodavatel: Apollo Scientific

PC302698-1G PC302698-5G
APOSPC302698-1GEA 4770 CZK
APOSPC302698-1G APOSPC302698-5G
2-Methyl-5-(trifluoromethyl)phenol 98%
2-Methyl-5-(trifluoromethyl)phenol
Vzorec: C₈H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD09832287
CAS číslo: 1017778-05-0
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR