2-Fluoro-5-toluoyl chloride
Dodavatel: Apollo Scientific
PC302689-10G
PC302689-5G
APOSPC302689-10GEA
9330
CZK
APOSPC302689-10G
APOSPC302689-5G
2-Fluoro-5-toluoyl chloride
2-Fluoro-5-toluoyl chloride
Vzorec:
C₈H₆ClFO Storage Temperature: Ambient |
MDL Number:
MFCD04973776 CAS číslo: 135564-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...