2-Fluoro-5-methyl-4-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302687-1G
PC302687-5G
APOSPC302687-1GEA
8900
CZK
APOSPC302687-1G
APOSPC302687-5G
2-Fluoro-5-methyl-4-(trifluoromethyl)benzonitrile 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₅F₄N MW: 203.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687195 CAS číslo: 1323966-20-6 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...