Tisk…

2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde 98%

Dodavatel: Apollo Scientific

PC302681-10G PC302681-5G PC302681-1G
APOSPC302681-10GEA 7480 CZK
APOSPC302681-10G APOSPC302681-5G APOSPC302681-1G
2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde 98%
2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde
Vzorec: C₉H₃F₇O
MW: 260,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258668
CAS číslo: 1017777-37-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR