(2-Ethoxy-3,5-difluorophenyl)acetic acid 98%
Dodavatel: Apollo Scientific
PC302680-1G
PC302680-5G
PC302680-10G
APOSPC302680-1GEA
4530
CZK
APOSPC302680-1G
APOSPC302680-5G
APOSPC302680-10G
(2-Ethoxy-3,5-difluorophenyl)acetic acid 98%
(2-Ethoxy-3,5-difluorophenyl)acetic acid
Vzorec:
C₁₀H₁₀F₂O₃ MW: 216,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258720 CAS číslo: 1017779-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...