Tisk…

2-Chloro-6-(trifluoromethyl)benzamide 98%

Dodavatel: Apollo Scientific

PC302675-5G PC302675-1G
APOSPC302675-5GEA 3870 CZK
APOSPC302675-5G APOSPC302675-1G
2-Chloro-6-(trifluoromethyl)benzamide 98%
2-Chloro-6-(trifluoromethyl)benzamide
Vzorec: C₈H₅ClF₃NO
MW: 223,58 g/mol
Bod tání: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD09025362
CAS číslo: 916420-45-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR