Tisk…

2-Chloro-6-fluoro-3-methylaniline 98%

Dodavatel: Apollo Scientific

PC302641-10G PC302641-5G PC302641-1G
APOSPC302641-10GEA 6390 CZK
APOSPC302641-10G APOSPC302641-5G APOSPC302641-1G
2-Chloro-6-fluoro-3-methylaniline 98%
2-Chloro-6-fluoro-3-methylaniline
Vzorec: C₇H₇ClFN
MW: 159,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660170
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR