Tisk…

6-Chloro-2,3-difluorobenzylamine 98%

Dodavatel: Apollo Scientific

Synonyms: 6-Chloro-2,3-difluorobenzylamine

PC302629-1G PC302629-5G
APOSPC302629-1GEA 3310 CZK
APOSPC302629-1G APOSPC302629-5G
6-Chloro-2,3-difluorobenzylamine 98%
6-Chloro-2,3-difluorobenzylamine
Vzorec: C₇H₆ClF₂N
MW: 177,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201055
CAS číslo: 1373921-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR