Tisk…

6-Chloro-2,3-difluorobenzaldehyde 97%

Dodavatel: Apollo Scientific

PC302627-25G PC302627-5G
APOSPC302627-25GEA 6350 CZK
APOSPC302627-25G APOSPC302627-5G
6-Chloro-2,3-difluorobenzaldehyde 97%
6-Chloro-2,3-difluorobenzaldehyde
Vzorec: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD07368118
CAS číslo: 797791-33-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR