Tisk…

5-Chloro-2-(trifluoromethoxy)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302614-1G PC302614-5G
APOSPC302614-1GEA 5060 CZK
APOSPC302614-1G APOSPC302614-5G
5-Chloro-2-(trifluoromethoxy)benzyl bromide 98%
5-Chloro-2-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅BrClF₃O
MW: 289.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519358
CAS číslo: 1092461-19-2
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR