Tisk…

5-(Difluoromethoxy)-3-fluorobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC302611-1G PC302611-5G
APOSPC302611-1GEA 3310 CZK
APOSPC302611-1G APOSPC302611-5G
5-(Difluoromethoxy)-3-fluorobenzyl bromide 95%
5-(Difluoromethoxy)-3-fluorobenzyl bromide
Vzorec: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832362
CAS číslo: 1017779-39-3
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR