Tisk…

4-Methyl-3-(trifluoromethyl)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC302609-1G PC302609-5G PC302609-10G
APOSPC302609-1GEA 5130 CZK
APOSPC302609-1G APOSPC302609-5G APOSPC302609-10G
4-Methyl-3-(trifluoromethyl)phenylacetonitrile 97%
4-Methyl-3-(trifluoromethyl)phenylacetonitrile
Vzorec: C₁₀H₈F₃N
Storage Temperature: Ambient
MDL Number: MFCD09832291
CAS číslo: 1000526-63-5
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR