Tisk…

4-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid

PC302597-10G PC302597-5G
APOSPC302597-10GEA 13630 CZK
APOSPC302597-10G APOSPC302597-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid
Vzorec: C₉H₆F₄O₂
MW: 222,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201064
CAS číslo: 1373920-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR