Tisk…

4-Ethoxy-3,5-difluorobenzylamine 97%

Dodavatel: Apollo Scientific

PC302588-1G PC302588-5G
APOSPC302588-1GEA 3870 CZK
APOSPC302588-1G APOSPC302588-5G
4-Ethoxy-3,5-difluorobenzylamine 97%
4-Ethoxy-3,5-difluorobenzylamine
Vzorec: C₉H₁₁F₂NO
MW: 187,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258689
CAS číslo: 1017779-32-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR