4-Chloro-3,5-difluorobenzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC302574-10G
PC302574-5G
PC302574-1G
PC302574-25G
APOSPC302574-10GEA
16570
CZK
APOSPC302574-10G
APOSPC302574-5G
APOSPC302574-1G
APOSPC302574-25G
4-Chloro-3,5-difluorobenzaldehyde 97%
4-Chloro-3,5-difluorobenzaldehyde
Vzorec:
C₇H₃ClF₂O Storage Temperature: Ambient |
MDL Number:
MFCD11553555 CAS číslo: 1160573-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...