4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302573-25G
PC302573-5G
PC302573-50G
APOSPC302573-25GEA
12800
CZK
APOSPC302573-25G
APOSPC302573-5G
APOSPC302573-50G
4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%
4-Chloro-3-(trifluoromethylthio)benzyl bromide
Vzorec:
C₈H₅BrClF₃S Storage Temperature: Ambient |
MDL Number:
MFCD06660314 CAS číslo: 886503-38-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...