Tisk…

4-Chloro-2-fluoro-3-methoxybenzonitrile 97%

Dodavatel: Apollo Scientific

PC302570-1G PC302570-5G
APOSPC302570-1GEA 3150 CZK
APOSPC302570-1G APOSPC302570-5G
4-Chloro-2-fluoro-3-methoxybenzonitrile 97%
4-Chloro-2-fluoro-3-methoxybenzonitrile
Vzorec: C₈H₅ClFNO
MW: 185,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687208
CAS číslo: 1126320-68-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR