Tisk…

4-Chloro-2-fluoro-3-methoxybenzamide 98%

Dodavatel: Apollo Scientific

Synonyms: 4-chloro-2-fluoro-3-methoxybenzamide

PC302569-1G PC302569-5G
APOSPC302569-1GEA 3310 CZK
APOSPC302569-1G APOSPC302569-5G
4-Chloro-2-fluoro-3-methoxybenzamide 98%
4-Chloro-2-fluoro-3-methoxybenzamide
Vzorec: C₈H₇ClFNO₂
MW: 203,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687206
CAS číslo: 1323966-12-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR