Tisk…

4-Chloro-2-(trifluoromethoxy)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC302566-1G PC302566-5G PC302566-10G
APOSPC302566-1GEA 3310 CZK
APOSPC302566-1G APOSPC302566-5G APOSPC302566-10G
4-Chloro-2-(trifluoromethoxy)benzyl bromide 97%
4-Chloro-2-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD18393833
CAS číslo: 1261606-06-7
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR