3'-Fluoro-5'-methylacetophenone 98%
Dodavatel: Apollo Scientific
PC302555-10G
PC302555-5G
PC302555-1G
PC302555-100MG
APOSPC302555-10GEA
5820
CZK
APOSPC302555-10G
APOSPC302555-5G
APOSPC302555-1G
APOSPC302555-100MG
3'-Fluoro-5'-methylacetophenone 98%
3'-Fluoro-5'-methylacetophenone
Vzorec:
C₉H₉FO MW: 152,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115883 CAS číslo: 886497-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...