(3-Ethoxy-2,4-difluorophenyl)acetic acid 98%
Dodavatel: Apollo Scientific
PC302545-1G
PC302545-5G
APOSPC302545-1GEA
3870
CZK
APOSPC302545-1G
APOSPC302545-5G
(3-Ethoxy-2,4-difluorophenyl)acetic acid 98%
(3-Ethoxy-2,4-difluorophenyl)acetic acid
Vzorec:
C₁₀H₁₀F₂O₃ Bod tání: 104…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258731 CAS číslo: 1017778-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...