Tisk…

3-Chloro-5-(pentafluorothio)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC302530-1G PC302530-5G PC302530-10G
APOSPC302530-1GEA 9920 CZK
APOSPC302530-1G APOSPC302530-5G APOSPC302530-10G
3-Chloro-5-(pentafluorothio)benzyl bromide 97%
3-Chloro-5-(pentafluorothio)benzyl bromide
Vzorec: C₇H₅BrClF₅S
MW: 331,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652423
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR