Tisk…

3,6-Dichloro-2-fluorobenzamide 97%

Dodavatel: Apollo Scientific

PC302495-5G PC302495-10G PC302495-1G
APOSPC302495-5GEA 3870 CZK
APOSPC302495-5G APOSPC302495-10G APOSPC302495-1G
3,6-Dichloro-2-fluorobenzamide 97%
3,6-Dichloro-2-fluorobenzamide
Vzorec: C₇H₄Cl₂FNO
Storage Temperature: Ambient
MDL Number: MFCD09025384
CAS číslo: 916420-64-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR