3-(3-Fluoro-5-methylphenyl)propionic acid 98%
Dodavatel: Apollo Scientific
PC302484-1G
PC302484-5G
PC302484-10G
APOSPC302484-1GEA
3310
CZK
APOSPC302484-1G
APOSPC302484-5G
APOSPC302484-10G
3-(3-Fluoro-5-methylphenyl)propionic acid 98%
3-(3-Fluoro-5-methylphenyl)propionic acid
Vzorec:
C₁₀H₁₁FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09832369 CAS číslo: 1017779-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...