3-(3-Fluoro-2-methylphenyl)propionic acid 98%
Dodavatel: Apollo Scientific
PC302483-1G
PC302483-5G
APOSPC302483-1GEA
3310
CZK
APOSPC302483-1G
APOSPC302483-5G
3-(3-Fluoro-2-methylphenyl)propionic acid 98%
3-(3-Fluoro-2-methylphenyl)propionic acid
Vzorec:
C₁₀H₁₁FO₂ Storage Temperature: Ambient |
CAS číslo:
377083-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...