3-(3-Fluoro-2-methoxyphenyl)propionic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
3-(3-Fluoro-2-methoxyphenyl)propanoic acid
PC302482-1G
PC302482-5G
PC302482-10G
APOSPC302482-1GEA
2780
CZK
APOSPC302482-1G
APOSPC302482-5G
APOSPC302482-10G
3-(3-Fluoro-2-methoxyphenyl)propionic acid 97%
3-(3-Fluoro-2-methoxyphenyl)propionic acid
Vzorec:
C₁₀H₁₁FO₃ MW: 198,1900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09999420 CAS číslo: 1092460-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...