2-Methyl-5-(trifluoromethoxy)benzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302472-5G
PC302472-1G
APOSPC302472-5GEA
8240
CZK
APOSPC302472-5G
APOSPC302472-1G
2-Methyl-5-(trifluoromethoxy)benzaldehyde 98%
2-Methyl-5-(trifluoromethoxy)benzaldehyde
Vzorec:
C₉H₇F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18399706 CAS číslo: 195622-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...