Tisk…

2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide 98%

Dodavatel: Apollo Scientific

Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzamide

PC302459-1G PC302459-5G
APOSPC302459-1GEA 8070 CZK
APOSPC302459-1G APOSPC302459-5G
2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide
Vzorec: C₉H₇F₄NO
MW: 221,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687194
CAS číslo: 1323966-43-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR