Tisk…

2-Fluoro-4-(trifluoromethoxy)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-2-fluoro-4-(trifluoromethoxy)benzene

PC302454-1G PC302454-5G PC302454-10G
APOSPC302454-1GEA 11080 CZK
APOSPC302454-1G APOSPC302454-5G APOSPC302454-10G
2-Fluoro-4-(trifluoromethoxy)benzyl bromide 98%
2-Fluoro-4-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD16652442
CAS číslo: 1240257-47-9
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR