2-Chloro-6-fluoro-3-methylbenzonitrile 98%
Dodavatel: Apollo Scientific
PC302448-25G
PC302448-5G
PC302448-1G
APOSPC302448-25GEA
12800
CZK
APOSPC302448-25G
APOSPC302448-5G
APOSPC302448-1G
2-Chloro-6-fluoro-3-methylbenzonitrile 98%
2-Chloro-6-fluoro-3-methylbenzonitrile
Vzorec:
C₈H₅ClFN MW: 169,59 g/mol Bod tání: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660169 CAS číslo: 886500-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...