α-Bromo-2,3-dichloro-6-fluorotoluene 98%
Dodavatel: Apollo Scientific
PC302429-10G
PC302429-5G
PC302429-50G
APOSPC302429-10GEA
10580
CZK
APOSPC302429-10G
APOSPC302429-5G
APOSPC302429-50G
α-Bromo-2,3-dichloro-6-fluorotoluene 98%
α-Bromo-2,3-dichloro-6-fluorotoluene
Vzorec:
C₇H₄BrCl₂F MW: 257,92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115873 CAS číslo: 886497-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...