Tisk…

2,3-Dichloro-6-fluorobenzonitrile 98%

Dodavatel: Apollo Scientific

PC302428-5G PC302428-1G
APOSPC302428-5GEA 5790 CZK
APOSPC302428-5G APOSPC302428-1G
2,3-Dichloro-6-fluorobenzonitrile 98%
2,3-Dichloro-6-fluorobenzonitrile
Vzorec: C₇H₂Cl₂FN
Bod tání: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD04115878
CAS číslo: 79544-26-6
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR