Tisk…

2,3-Dichloro-6-(trifluoromethyl)phenylacetic acid 97%

Dodavatel: Apollo Scientific
Varování

PC302427-1G PC302427-5G
APOSPC302427-1GEA 4960 CZK
APOSPC302427-1G APOSPC302427-5G
2,3-Dichloro-6-(trifluoromethyl)phenylacetic acid 97%
2,3-Dichloro-6-(trifluoromethyl)phenylacetic acid
Vzorec: C₉H₅Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09832274
CAS číslo: 1017777-86-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR