Tisk…

2-(Difluoromethoxy)-4-fluorobenzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302419-1G PC302419-5G PC302419-10G
APOSPC302419-1GEA 3640 CZK
APOSPC302419-1G APOSPC302419-5G APOSPC302419-10G
2-(Difluoromethoxy)-4-fluorobenzyl bromide 98%
2-(Difluoromethoxy)-4-fluorobenzyl bromide
Vzorec: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832363
CAS číslo: 1017779-42-8
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR