5-Chloro-2-(trifluoromethyl)benzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302415-25G
PC302415-5G
PC302415-10G
APOSPC302415-25GEA
6050
CZK
APOSPC302415-25G
APOSPC302415-5G
APOSPC302415-10G
5-Chloro-2-(trifluoromethyl)benzaldehyde 98%
5-Chloro-2-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₄ClF₃O Storage Temperature: Ambient |
MDL Number:
MFCD03412204 CAS číslo: 90381-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...