Tisk…

(4,5-Difluoro-2-methoxyphenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4,5-Difluoro-2-methoxyphenyl)acetic acid

PC302405-1G PC302405-5G PC302405-250MG
APOSPC302405-1GEA 10550 CZK
APOSPC302405-1G APOSPC302405-5G APOSPC302405-250MG
(4,5-Difluoro-2-methoxyphenyl)acetic acid 98%
(4,5-Difluoro-2-methoxyphenyl)acetic acid
Vzorec: (C₆H₂F₂(OCH₃))CH₂COOH
MW: 186,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660348
CAS číslo: 886761-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR