Tisk…

3-Chloro-5-(trifluoromethyl)benzyl alcohol 95%

Dodavatel: Apollo Scientific

PC302399-10G PC302399-5G PC302399-1G
APOSPC302399-10GEA 6750 CZK
APOSPC302399-10G APOSPC302399-5G APOSPC302399-1G
3-Chloro-5-(trifluoromethyl)benzyl alcohol 95%
3-Chloro-5-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115848
CAS číslo: 886496-87-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR