Tisk…

(2,3-Difluoro-4-methoxyphenyl)acetic acid 98%

Dodavatel: Apollo Scientific

PC302373-1G PC302373-5G PC302373-25G
APOSPC302373-1GEA 2650 CZK
APOSPC302373-1G APOSPC302373-5G APOSPC302373-25G
(2,3-Difluoro-4-methoxyphenyl)acetic acid 98%
(2,3-Difluoro-4-methoxyphenyl)acetic acid
Vzorec: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06660319
CAS číslo: 886503-70-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR