Tisk…

4-Methoxy-3-(trifluoromethoxy)benzoyl chloride 98%

Dodavatel: Apollo Scientific

PC302345-1G PC302345-5G
APOSPC302345-1GEA 4800 CZK
APOSPC302345-1G APOSPC302345-5G
4-Methoxy-3-(trifluoromethoxy)benzoyl chloride 98%
4-Methoxy-3-(trifluoromethoxy)benzoyl chloride
Vzorec: C₉H₆ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18398465
CAS číslo: 1070774-23-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR