Tisk…

4-Fluoro-2-(trifluoromethoxy)benzyl alcohol

Dodavatel: Apollo Scientific

PC302339-1G PC302339-5G
APOSPC302339-1GEA 5130 CZK
APOSPC302339-1G APOSPC302339-5G
4-Fluoro-2-(trifluoromethoxy)benzyl alcohol
4-Fluoro-2-(trifluoromethoxy)benzyl alcohol
Vzorec: C₈H₆F₄O₂
MW: 210,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687174
CAS číslo: 1242252-68-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR