Tisk…

4-Fluoro-2-(trifluoromethoxy)benzoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-fluoro-2-(trifluoromethoxy)benzoyl chloride

PC302338-1G PC302338-5G
APOSPC302338-1GEA 5130 CZK
APOSPC302338-1G APOSPC302338-5G
4-Fluoro-2-(trifluoromethoxy)benzoyl chloride
4-Fluoro-2-(trifluoromethoxy)benzoyl chloride
Vzorec: C₈H₃ClF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD19687189
CAS číslo: 1323966-38-6
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR