4-Ethoxy-3-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
PC302335-5G
PC302335-10G
PC302335-1G
APOSPC302335-5GEA
4200
CZK
APOSPC302335-5G
APOSPC302335-10G
APOSPC302335-1G
4-Ethoxy-3-(trifluoromethyl)benzonitrile 98%
4-Ethoxy-3-(trifluoromethyl)benzonitrile
Vzorec:
C₁₀H₈F₃NO MW: 215.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13196579 CAS číslo: 1206593-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...