4-Ethoxy-2,6-difluorobenzyl bromide 98%
Dodavatel: Apollo Scientific
PC302331-1G
PC302331-5G
PC302331-25G
APOSPC302331-1GEA
0
CZK
APOSPC302331-1G
APOSPC302331-5G
APOSPC302331-25G
4-Ethoxy-2,6-difluorobenzyl bromide 98%
4-Ethoxy-2,6-difluorobenzyl bromide
Vzorec:
C₉H₉BrF₂O MW: 251,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258697 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...