Tisk…

4-Chloro-2-(trifluoromethoxy)benzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

PC302320-1G PC302320-5G
APOSPC302320-1GEA 2650 CZK
APOSPC302320-1G APOSPC302320-5G
4-Chloro-2-(trifluoromethoxy)benzyl alcohol 98%
4-Chloro-2-(trifluoromethoxy)benzyl alcohol
Vzorec: C₈H₆ClF₃O₂
MW: 226,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393819
CAS číslo: 1261673-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR