4-Chloro-2-(trifluoromethoxy)benzaldehyde 97%
Dodavatel: Apollo Scientific
PC302319-1G
PC302319-5G
PC302319-10G
APOSPC302319-1GEA
1040
CZK
APOSPC302319-1G
APOSPC302319-5G
APOSPC302319-10G
4-Chloro-2-(trifluoromethoxy)benzaldehyde 97%
4-Chloro-2-(trifluoromethoxy)benzaldehyde
Vzorec:
C₈H₄ClF₃O₂ MW: 224,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18393772 CAS číslo: 1261442-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...