4-(Difluoromethoxy)-2-fluorobenzonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC302314-1G
PC302314-5G
APOSPC302314-1GEA
2660
CZK
APOSPC302314-1G
APOSPC302314-5G
4-(Difluoromethoxy)-2-fluorobenzonitrile 97%
4-(Difluoromethoxy)-2-fluorobenzonitrile
Vzorec:
C₈H₄F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD09832366 CAS číslo: 1017779-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...