3-Methoxy-5-(trifluoromethyl)phenol 98%
Dodavatel: Apollo Scientific
PC302309-1G
PC302309-5G
PC302309-25G
APOSPC302309-1GEA
1240
CZK
APOSPC302309-1G
APOSPC302309-5G
APOSPC302309-25G
3-Methoxy-5-(trifluoromethyl)phenol 98%
3-Methoxy-5-(trifluoromethyl)phenol
Vzorec:
C₈H₇F₃O₂ Bod tání: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD09025418 CAS číslo: 349-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...