3-Fluoro-5-(trifluoromethoxy)benzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302305-1G
PC302305-5G
APOSPC302305-1GEA
11150
CZK
APOSPC302305-1G
APOSPC302305-5G
3-Fluoro-5-(trifluoromethoxy)benzylamine 98%
3-Fluoro-5-(trifluoromethoxy)benzylamine
Vzorec:
C₈H₇F₄NO MW: 209,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18783229 CAS číslo: 1353000-19-7 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...