Tisk…

3-Fluoro-5-(trifluoromethoxy)benzoyl chloride

Dodavatel: Apollo Scientific

Synonyms: 3-fluoro-5-(trifluoromethoxy)benzoyl chloride

PC302303-1G PC302303-5G
APOSPC302303-1GEA 16570 CZK
APOSPC302303-1G APOSPC302303-5G
3-Fluoro-5-(trifluoromethoxy)benzoyl chloride
3-Fluoro-5-(trifluoromethoxy)benzoyl chloride
Vzorec: C₈H₃ClF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18783222
CAS číslo: 1352999-78-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR